Learn More
Glyoxylic acid, 50% w/w aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B251490B
Description
Glyoxylic acid is used in Hopkins Cole reaction, which is used in the detection of tryptophan in proteins. It reacts with phenol to get 4-hydroxymandelic acid, which on further reaction with ammonia gives hydroxyphenylglycine, as a precursor to the drug amoxicillin. It is also used as a starting material for the preparation of 4-hydroxyphenylacetic acid, which is used to get atenolol. It is involved in the production of agrochemicals, aromas, cosmetic ingredient and pharmaceutical intermediate. It is also used in water purification and in the preservation of food. Further, it is employed as precursor in the synthesis of iron chelates. In addition to this, it serves as an intermediate of varnish material and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlyoxylic acid is used in Hopkins Cole reaction, which is used in the detection of tryptophan in proteins. It reacts with phenol to get 4-hydroxymandelic acid, which on further reaction with ammonia gives hydroxyphenylglycine, as a precursor to the drug amoxicillin. It is also used as a starting material for the preparation of 4-hydroxyphenylacetic acid, which is used to get atenolol. It is involved in the production of agrochemicals, aromas, cosmetic ingredient and pharmaceutical intermediate. It is also used in water purification and in the preservation of food. Further, it is employed as precursor in the synthesis of iron chelates. In addition to this, it serves as an intermediate of varnish material and dyes.
Solubility
Miscible with ethanol. Slightly miscible with ether and benzene. Immiscible with esters.
Notes
Incompatible with metals, alkalies, strong oxidizing agents and strong bases.
Specifications
Glyoxylic acid | |
-93°C | |
111°C | |
MFCD00006958 | |
UN3265 | |
Miscible with ethanol. Slightly miscible with ether and benzene. Immiscible with esters. | |
C(=O)C(=O)O | |
74.035 | |
CHEBI:16891 | |
50% w/w aq. soln. |
298-12-4 | |
1.339 | |
C2H2O3 | |
1000 g | |
glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid | |
HHLFWLYXYJOTON-UHFFFAOYSA-N | |
oxaldehydic acid | |
760 | |
74.04 |
Chemical Identifiers
298-12-4 | |
74.035 | |
HHLFWLYXYJOTON-UHFFFAOYSA-N | |
760 | |
oxaldehydic acid |
C2H2O3 | |
MFCD00006958 | |
glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid | |
CHEBI:16891 | |
C(=O)C(=O)O |
Safety and Handling
GHS H Statement
H314-H317
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
P234-P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H290-H314-H317-H318
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-058-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only