missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glyoxal sodium bisulfite hydrate, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 332360-05-1 |
---|---|
Molecular Formula | C2H4Na2O8S2·xH2O |
Molecular Weight (g/mol) | 266.16 |
InChI Key | JQYGKJGYGOFFIK-UHFFFAOYSA-L |
Synonym | glyoxal-sodium bisulfite monohydrate, glyoxal sodium bisulfite addition compound hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt, c2h4o8s2.2na.h2o, glyoxal-sodium bisulfite monohydrate mi, glyoxal bis sodium hydrogen sulfite adduct hydrate, glyoxal sodium bisulfite addition compound hydrate,98, disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate, disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate |
PubChem CID | 16211366 |
IUPAC Name | disodium;1,2-dihydroxyethane-1,2-disulfonate;hydrate |
SMILES | C(C(O)S(=O)(=O)[O-])(O)S(=O)(=O)[O-].O.[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC173270250
|
Thermo Scientific Chemicals
173270250 |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
332360-05-1 | |
266.16 | |
glyoxal-sodium bisulfite monohydrate, glyoxal sodium bisulfite addition compound hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt, c2h4o8s2.2na.h2o, glyoxal-sodium bisulfite monohydrate mi, glyoxal bis sodium hydrogen sulfite adduct hydrate, glyoxal sodium bisulfite addition compound hydrate,98, disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate, disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate | |
disodium;1,2-dihydroxyethane-1,2-disulfonate;hydrate |
C2H4Na2O8S2·xH2O | |
JQYGKJGYGOFFIK-UHFFFAOYSA-L | |
16211366 | |
C(C(O)S(=O)(=O)[O-])(O)S(=O)(=O)[O-].O.[Na+].[Na+] |
Specifications
332360-05-1 | |
97.5% min | |
[-CH(OH)SO3Na]2·xH2O | |
glyoxal-sodium bisulfite monohydrate, glyoxal sodium bisulfite addition compound hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt, c2h4o8s2.2na.h2o, glyoxal-sodium bisulfite monohydrate mi, glyoxal bis sodium hydrogen sulfite adduct hydrate, glyoxal sodium bisulfite addition compound hydrate,98, disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate, disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate | |
JQYGKJGYGOFFIK-UHFFFAOYSA-L | |
disodium;1,2-dihydroxyethane-1,2-disulfonate;hydrate | |
16211366 | |
98% | |
10% max. (120°C, 2 hrs) |
25g | |
C2H4Na2O8S2·xH2O | |
14,451 | |
Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol | |
C(C(O)S(=O)(=O)[O-])(O)S(=O)(=O)[O-].O.[Na+].[Na+] | |
266.16 | |
266.16 | |
Authentic | |
Glyoxal sodium bisulfite hydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO â Research Use Only