missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glyoxal, pure, 40 wt.% solution in water

CAS: 107-22-2 | C2H2O2 | 58.04 g/mol

Supplier:  Thermo Scientific Chemicals 156220010

 View more versions of this product

Catalog No. AC156220010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glyoxal
107-22-2,7732-18-5
Colorless to Yellow
104.0°C
Glass bottle
HCOCHO
1 L
01,413
15, 4544
Solubility in water: miscible.
C(=O)C=O
58.04
8 mPa.s (20°C)
58.04
Pure
40 wt.% Solution in Water
-14.0°C
1.2650g/mL
>104°C
C2H2O2
MFCD00006957
01, 759
1.265
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
oxaldehyde
7860
CHEBI:34779
39 to 41% (Titrimetry other)
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only