missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glyoxal, 40% w/w aq. soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A16144AP

 View more versions of this product

Catalog No. AAA16144AP



Description

Description

Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal is used to prepare 4,5-dihydroxy-2-imidazolidinone by condensation with urea. It finds application in leather tanning process, textile finishes and paper coatings. It is an important building block in the synthesis of imidazoles. It acts as a solubilizer and cross-linking agent in polymer chemistry. Further, it is used as a fixative for histology to preserve cells in order to examine under a microscope.

Solubility
Miscible with water, methanol and dimethylsulfoxide.

Notes
Moisture and heat sensitive. Incompatible with strong oxidizing agents, strong reducing agent, oxygen, peroxides, amides, amines, hydroxy-containing materials, nitric acid and aldehydes.
Specifications

Specifications

Glyoxal
-14°C
104°C
1.409
MFCD00006957
1732463
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
oxaldehyde
7860
58.04
107-22-2
1.271
C2H2O2
HCOCHO
500 mL
14,4509
Miscible with water,methanol and dimethylsulfoxide.
C(=O)C=O
58.036
CHEBI:34779
40% w/w aq. soln.

Chemical Identifiers

107-22-2
58.036
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
7860
oxaldehyde
C2H2O2
MFCD00006957
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
CHEBI:34779
C(=O)C=O
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H317
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c

H315-H317-H319-H332-H335-H341

EINECSNumber : 203-474-9

RTECSNumber : MD2700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only