missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycyrrhizic acid monoammonium salt, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 213751000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Glycyrrhizic acid monoammonium salt | |
53956-04-0 | |
98+% | |
C42H65NO16 | |
100g | |
+ 51.50 (20.00°C c=1,EtOH 50% v/v) | |
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid, ethanol,, soluble in ammonia water | |
N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O | |
839.97 | |
839.98 | |
10% max. 18-alpha-glycyrrhizic acid (HPLC) |
98% | |
Authentic | |
Glass bottle | |
MFCD00167400,MFCD00167400 | |
10,4369 | |
+ 51.50 | |
ILRKKHJEINIICQ-OOFFSTKBSA-N | |
(2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(4aR,6aR,6bS,8aS,11R,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carb | |
131676200 | |
98+% |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 258-887-7
RTECSNumber : LZ6500000
TSCA : TSCA
RUO â Research Use Only