Please login to your online account to display your discounted pricing

Glycyrrhizic acid monoammonium salt, 98+%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC213750100

 View more versions of this product

Catalog No. AC213750100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 839.98
Physical Form Powder or Crystals
Quantity 10g
Color Yellow to yellow-green
Assay 98.0 to 102.0% (anhydrous substance) (HClO4 titr.) (Total base)
TSCA TSCA
RTECS Number LZ6500000
EINECS Number 258-887-7
Specific Rotation Condition + 51.50 (20.00°C c=1,EtOH 50% v/v)
Related Substances 10.0% max. 18 alpha−glycyrrhizic acid (HPLC)
Health Hazard 1 HYGROSCOPIC
Heavy Metals 20ppm max.
Sulfated Ash 0.2% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Water 7.5% max. (K.F.)
Specific Rotation +49.0° to +54.0° (20°C, 589nm) (c=1, EtOH 50% v/v) (anhydrous substance)
Assay Percent Range 98+%
Name Note 98%
Molecular Formula C42H65NO16
CAS 53956-04-0
Chemical Name or Material Glycyrrhizic acid monoammonium salt
MDL Number MFCD00150156

Specifications

Chemical Name or Material Glycyrrhizic acid monoammonium salt
Name Note 98%
CAS 53956-04-0
Assay 98.0 to 102.0% (anhydrous substance) (HClO4 titr.) (Total base)
Assay Percent Range 98+%
Molecular Formula C42H65NO16
Formula Weight 839.98
MDL Number MFCD00150156
Accessories

HYGROSCOPIC

SDS & Safety Documents