missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycylglycine, 0.2M buffer soln., pH 7.5

$84.96 - $133.08

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60282AE View Documents Alfa Aesar™
J60282AE
100mL Each for $84.96
 
AAJ60282AK View Documents Alfa Aesar™
J60282AK
250mL Each for $133.08
 
Specifications

Chemical Identifiers

556-50-3
132.119
YMAWOPBAYDPSLA-UHFFFAOYSA-N
CHEBI:17201
C(C(=O)NCC(=O)O)N
C4H8N2O3
MFCD00008130
11163
2-[(2-aminoacetyl)amino]acetic acid

Specifications

556-50-3
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
2-[(2-aminoacetyl)amino]acetic acid
11163
Liquid
7.5
100mL
C4H8N2O3
0.2M buffer soln.
C(C(=O)NCC(=O)O)N
132.119
CHEBI:17201
Odorless
Colorless
Glycylglycine buffer soln.