Please login to your online account to display your discounted pricing

Glycyl-L-Glutamic Acid, Fisher BioReagents


Quantity 100mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2522100

 View more versions of this product

Catalog No. BP2522100

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 155°C
Formula Weight 204.18
Quantity 100mg
Physical Form Solid
Molecular Formula C7H12N2O5
Synonym Gly-Glu
CAS 7412-78-4
Health Hazard 2 CAUTION!
Chemical Name or Material Glycyl-L-Glutamic Acid
Packaging Amber Glass
ChemAlert Storage Symbol Gray
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Recommended Storage 0°C
Color Beige

Specifications

Chemical Name or Material Glycyl-L-Glutamic Acid
CAS 7412-78-4
Physical Form Solid
Molecular Formula C7H12N2O5
Formula Weight 204.18
Synonym Gly-Glu
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

0°C

SDS & Safety Documents