missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycyl diphenylborinate, 95%

Manufacturer:  Alfa Aesar™ H5544303

 View more versions of this product

Catalog No. AAH5544303



Specifications

Specifications

Glycyl diphenylborinate
14335-29-6
MFCD05863277
IVHLNYVPKWICDY-UHFFFAOYSA-N
diphenylboranyl 2-aminoacetate
11840443
95%
1g
C14H14BNO2
2-diphenylboryl oxy-2-oxoethanamine
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)CN
239.081
239.08

Chemical Identifiers

14335-29-6
239.081
IVHLNYVPKWICDY-UHFFFAOYSA-N
11840443
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)CN
C14H14BNO2
MFCD05863277
2-diphenylboryl oxy-2-oxoethanamine
diphenylboranyl 2-aminoacetate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents