missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycosylated Peptide Certified Mass Spec Standard Mixture I

For cellular and molecular biology applications

Specifications

Product Type Glycosylated Peptide Certified Mass Spec Standard Mixture I, 500pmol each peptide per vial
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67704LPL Alfa Aesar™
J67704LPL
1 vial, 500 pmol N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67704ODL Alfa Aesar™
J67704ODL
3 vials, 500 pmol/vial N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

Glycosylated Peptide Certified Mass Spec Standard Mixture I, 500pmol each peptide per vial
Lyophilized powder
HPLC
-30° to -10°C
RUO