Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycosylated Peptide Certified Mass Spec Standard Mixture I


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67704LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67704LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial, 500 pmol
Format Lyophilized powder
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Glycosylated Peptide Certified Mass Spec Standard Mixture I, 500pmol each peptide per vial 1 vial
Purification Method HPLC