Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycosylated MUC5AC 3, Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67821ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67821ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 3 vials, 500 pmol/vial
For Use With (Application) For use in developing methods for glycopeptide mapping. Also used in standardization of affinity capture and selective enrichment protocols
Format Lyophilized powder
Sequence GT-T(GalNAc)-PSPVPTTSTTSAP
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Glycosylated MUC5AC 3, Certified Mass Spec Grade
Purification Method HPLC