Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycosylated Carcinoembryonic Antigen CGM2 (176-186), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67804LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67804LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial, 500 pmol
Product Type Glycosylated Carcinoembryonic Antigen CGM2 (176-186), Certified Mass Spec Grade
Molecular Weight 1626.60
Sequence TYLWWVN-N(GlcNAc)-QSL
Purification Method HPLC
Format Lyophilized powder
Storage Requirements -30° to -10°C
Regulatory Status RUO