Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycosylated Carcinoembryonic Antigen CGM2 (176-186), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67804ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67804ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 3 vials, 500 pmol/vial
Format Lyophilized powder
Sequence TYLWWVN-N(GlcNAc)-QSL
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Glycosylated Carcinoembryonic Antigen CGM2 (176-186), Certified Mass Spec Grade
Molecular Weight 1626.60
Purification Method HPLC