Please login to your online account to display your discounted pricing

Thermo Scientific™ GlycoLink™ Immobilization Kits and Resin

Immobilize polyclonal antibodies or other glycoproteins, via periodate-oxidized sugars, onto hydrazide-activated polyacrylamide beads for AP, IP or pull-down assays.

$358.07 - $488.65

Specifications

Support Type Acrylamide
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    

PI53149

 
thermo scientific™
PI53149
UltraLink Hydrazide Resin Pack for $358.07

PI88942

 
thermo scientific™
88942
GlycoLink Micro Immobilization Kit Each for $488.65
Description & Specifications

Specifications

Support Type Acrylamide

Thermo Scientific™ GlycoLink Immobilization Kits provide affinity columns and reagents to immobilize glycoproteins through oxidized sugar groups for use in affinity-purification applications or studies of protein-protein interactions.

The GlycoLink Kits use hydrazide-activated resin and a special catalyst to prepare affinity columns with polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Any glycosylated proteins (even monoclonal antibodies) that contain periodate-oxidizable carbohydrate groups can be immobilized with these kits to make reusable affinity columns. Antibodies immobilized by this hydrazide immobilization chemistry have unobstructed antigen-binding sites and optimal purification capability. Prepared columns with stable glycoproteins or antibodies can be regenerated and reused at least five times for affinity-purification without significant loss in binding capacity.

Highlights:

  • High-capacity – immobilize 1 to 10mg of oxidized antibody or other glycoprotein per milliliter of resin

  • Fast and efficient – immobilize in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)

  • Specific immobilization – the hydrazide-activated UltraLink Resin conjugates only to purified glycoproteins whose sugar (e.g., sialic acid) groups have been gently oxidized with periodate

  • Stable attachment – resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilization step is required

  • Preserves antibody function – immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target

  • Convenient sizes – choose from two complete 10-column kits: one with 1mL columns and one with 0.1mL columns

UltraLink Hydrazide Resin:

  • Ligand: hydrazide
  • Support: Thermo Scientific UltraLink Biosupport
  • Spacer arm: 22 atoms
  • Loading: activated with at least 15μmol hydrazide per milliliter of settled resin
  • Format: 50% resin slurry in 0.02% sodium azide

Includes:

Kits contain GlycoLink Columns; coupling buffer, coupling catalyst and wash buffer; oxidizing reagent; desalting columns, PBS buffer pack; and column accessories

Recommended for:

Purify antigens using covalently coupled antibodies; Determine binding partners for a glycoprotein of interest; Perform large-scale affinity purifications