missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycolic acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics 154510250

 View more versions of this product

Catalog No. AC154510250



Specifications

Specifications

Glycolic acid
Vapor Pressure: 8.1mmHg at 80°C
C2H4O3
03, 228
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
C(C(=O)O)O
76.051
CHEBI:17497
Adhering Crystals
Conforms to Ref
1% max.
72.0°C to 80.0°C
100.0°C
>300°C
79-14-1
99%
HOCH2CO2H
15, 4534
AEMRFAOFKBGASW-UHFFFAOYSA-N
2-hydroxyacetic acid
757
76.04
99%
Glass Bottle
>100°C
White
25g

Chemical Identifiers

79-14-1
76.051
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
CHEBI:17497
C(C(=O)O)O
C2H4O3
AEMRFAOFKBGASW-UHFFFAOYSA-N
757
2-hydroxyacetic acid
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. Harmful if inhaled.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Danger

SDS Documents