missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycolic acid 70% in water, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 411032500

 View more versions of this product

Catalog No. AC411032500


 

Specifications Description & Specifications

Specifications

76.04
250g
65 to 69%
7732-18-5,79-14-1
Hydroxyacetic acid
C2H4O3
70% in water
Solution
Clear yellow
1.2700g/mL
Glass bottle
MFCD00004312
Glycolic acid
CH2OHCOOH

Chemical Identifiers

79-14-1,7732-18-5
CH2OHCOOH
MFCD00004312
Glycolic acid, 70% in water
C2H4O3
76.04
Hydroxyacetic acid
65 to 69%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. May damage fertility. May damage the unborn child.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Danger

EINECSNumber : 201-180-5

RTECSNumber : MC5250000

TSCA : TSCA