missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycol sulfite, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 120120250
Specifications
Glycol sulfite | |
79°C | |
Glass bottle | |
1.42 | |
Colorless | |
98% | |
98% | |
MFCD00005354 | |
WDXYVJKNSMILOQ-UHFFFAOYSA-N | |
1,3,2-dioxathiolane 2-oxide | |
77342 | |
Liquid |
1.4200g/mL | |
Authentic | |
1.4450 to 1.447 | |
68°C (20.0mmHg) | |
25g | |
3741-38-6 | |
C2H4O3S | |
1,2-ethylene sulfite, 1,3,2-dioxathiolane, 2-oxide, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulfite, ethylene sulphite, glycol sulfite, glycolsulfite, sulfurous acid, cyclic ester with ethylene glycol | |
C1COS(=O)O1 | |
108.11 | |
108.11 | |
98% |
Chemical Identifiers
3741-38-6 | |
108.11 | |
WDXYVJKNSMILOQ-UHFFFAOYSA-N | |
77342 | |
C1COS(=O)O1 |
C2H4O3S | |
MFCD00005354 | |
1,2-ethylene sulfite, 1,3,2-dioxathiolane, 2-oxide, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulfite, ethylene sulphite, glycol sulfite, glycolsulfite, sulfurous acid, cyclic ester with ethylene glycol | |
1,3,2-dioxathiolane 2-oxide |
Safety and Handling
EINECSNumber : 223-131-7
TSCA : TSCA