Please login to your online account to display your discounted pricing

Glycol Sulfite 98%, ACROS Organics™

$223.89 - $223.89

Chemical Identifiers

Chemical Name or Material Glycol sulfite
Name Note 98%
CAS 3741-38-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H4O3S
Formula Weight 108.11
MDL Number MFCD00005354
Synonym Ethylene sulfite
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC120120250

  SDS
fisher scientific acros
120120250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC120121000

  SDS
thermo fisher scientific
120121000
Glass bottle 100g Each for $223.89
Description & Specifications

Specifications

Formula Weight 108.11
Assay ≥97.5%
Physical Form Liquid
Quantity 25g
Color Clear colorless
TSCA TSCA
EINECS Number 223-131-7
Density 1.42g/mL
Packaging Glass bottle
Refractive Index 1.4450 to 1.4470 (20°C, 589nm)
Synonym Ethylene sulfite
Infrared Spectrum Authentic
Chemical Name or Material Glycol sulfite
MDL Number MFCD00005354
Assay Percent Range 98%
CAS 3741-38-6
Molecular Formula C2H4O3S
Name Note 98%