Please login to your online account to display your discounted pricing

Glycine, sodium salt hydrate, 99%, ACROS Organics™

$82.28 - $82.28

Chemical Identifiers

Chemical Name or Material Glycine, sodium salt
Name Note 99%
CAS 6000-44-8
Assay ≥98.5% (Total base)
Assay Percent Range 99%
Molecular Formula C2H4NNaO2
Linear Formula H2NCH2CO2Na
Formula Weight 97.05
MDL Number MFCD00150683
Synonym Sodium glycinate hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC204591000

  SDS
thermo fisher scientific
204591000
100g Each for $82.28

AC204595000

  SDS
thermo fisher scientific
AC204595000
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Moist Beads
Assay ≥98.5% (Total base)
Formula Weight 97.05
Quantity 100g
Color White to yellow
EINECS Number 227-842-3
TSCA TSCA
Water 1% Max. (K.F.)
pH 11.1 to 12.0 (10% aq. soln.)
RTECS Number MC1100000
Chemical Name or Material Glycine, sodium salt
Name Note 99%
Assay Percent Range 99%
Infrared Spectrum Authentic
Molecular Formula C2H4NNaO2
Linear Formula H2NCH2CO2Na
MDL Number MFCD00150683
CAS 6000-44-8
Synonym Sodium glycinate hydrate
Transmittance 94.0 % min. (c=5, H2O, 400nm) 1 cm cell