Please login to your online account to display your discounted pricing

Glycine, sodium salt hydrate, 99%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC204595000

 View more versions of this product

Catalog No. AC204595000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (Total base)
Formula Weight 97.05
Physical Form Moist Beads
Quantity 500g
Color White to yellow
TSCA TSCA
EINECS Number 227-842-3
pH 11.1 to 12.0 (10% aq. soln.)
RTECS Number MC1100000
Water 1% Max. (K.F.)
Infrared Spectrum Authentic
Transmittance 94.0 % min. (c=5, H2O, 400nm) 1 cm cell
Synonym Sodium glycinate hydrate
Assay Percent Range 99%
Linear Formula H2NCH2CO2Na
Molecular Formula C2H4NNaO2
MDL Number MFCD00150683
Chemical Name or Material Glycine, sodium salt
Name Note 99%
CAS 6000-44-8

Specifications

Chemical Name or Material Glycine, sodium salt
Name Note 99%
CAS 6000-44-8
Assay ≥98.5% (Total base)
Assay Percent Range 99%
Molecular Formula C2H4NNaO2
Linear Formula H2NCH2CO2Na
Formula Weight 97.05
MDL Number MFCD00150683
Synonym Sodium glycinate hydrate