Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycine n-octyl ester hydrochloride, 98%

$158.54 - $158.54

Chemical Identifiers

Chemical Name or Material Glycine n-octyl ester hydrochloride
CAS 39540-30-2
Molecular Formula H2NCH2CO2(CH2)7CH3·HCl
Formula Weight 223.75
MDL Number MFCD00151827
Beilstein 3696405
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1338506

 
alfa aesar™
L1338506
5g Each for $158.54
Description & Specifications

Specifications

Formula Weight 223.75
Chemical Name or Material Glycine n-octyl ester hydrochloride
Molecular Formula H2NCH2CO2(CH2)7CH3·HCl
MDL Number MFCD00151827
CAS 39540-30-2
Assay 98%
Beilstein 3696405
Melting Point 66° to 72°C
Quantity 5g