missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycine n-octyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ L1338503

 View more versions of this product

Catalog No. AAL1338503



Specifications

Specifications

Glycine n-octyl ester hydrochloride
C10H22ClNO2
3696405
OORFEZYGSYLFCL-UHFFFAOYSA-N
octyl 2-aminoacetate;hydrochloride
2759252
98%
1g
39540-30-2
MFCD00151827
octyl 2-aminoacetate hydrochloride, glycine n-octyl ester hydrochloride, glycine-1-octyl ester hydrochloride, pubchem12487, octyl2-aminoacetatehydrochloride, glycine octyl ester hydrochloride, octyl 2-azanylethanoate hydrochloride, 2-aminoacetic acid octyl ester hydrochloride
CCCCCCCCOC(=O)CN.Cl
223.741
223.75
66°C to 72°C

Chemical Identifiers

39540-30-2
223.741
OORFEZYGSYLFCL-UHFFFAOYSA-N
2759252
CCCCCCCCOC(=O)CN.Cl
C10H22ClNO2
MFCD00151827
octyl 2-aminoacetate hydrochloride, glycine n-octyl ester hydrochloride, glycine-1-octyl ester hydrochloride, pubchem12487, octyl2-aminoacetatehydrochloride, glycine octyl ester hydrochloride, octyl 2-azanylethanoate hydrochloride, 2-aminoacetic acid octyl ester hydrochloride
octyl 2-aminoacetate;hydrochloride
Documents