missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycine hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A121620E
Description
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Solubility
Soluble in water (>100 g/L).
Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
Specifications
175°C to 178°C | |
6000-43-7 | |
MFCD00012872 | |
14,4491 | |
Soluble in water (>100g/L). | |
[H+].[Cl-].NCC(O)=O | |
111.53 | |
111.53 | |
2500 g | |
Hygroscopic |
Glycine hydrochloride | |
C2H6ClNO2 | |
3909617 | |
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride | |
IVLXQGJVBGMLRR-UHFFFAOYSA-N | |
2-aminoacetic acid;hydrochloride | |
22316 | |
98% | |
Odorless |
Chemical Identifiers
6000-43-7 | |
111.53 | |
IVLXQGJVBGMLRR-UHFFFAOYSA-N | |
22316 | |
[H+].[Cl-].NCC(O)=O |
C2H6ClNO2 | |
MFCD00012872 | |
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride | |
2-aminoacetic acid;hydrochloride |
Safety and Handling
EINECSNumber : 227-841-8
RTECSNumber : MC0560000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only