missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ A1216222

 View more versions of this product

Catalog No. AAA1216222



Description

Description

Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Soluble in water (>100 g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
Specifications

Specifications

Glycine hydrochloride
C2H6ClNO2
3909617
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
C(C(=O)O)N.Cl
111.525
111.53
175°C to 178°C
Odorless
6000-43-7
MFCD00012872
14,4491
IVLXQGJVBGMLRR-UHFFFAOYSA-N
2-aminoacetic acid;hydrochloride
22316
98%
100g

Chemical Identifiers

6000-43-7
111.525
IVLXQGJVBGMLRR-UHFFFAOYSA-N
22316
C(C(=O)O)N.Cl
C2H6ClNO2
MFCD00012872
glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
2-aminoacetic acid;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : MC0560000

TSCA : TSCA

SDS Documents