Please login to your online account to display your discounted pricing

Glycine, For Ozone, 7% (w/v), LabChem™

$73.23 - $73.23

Chemical Identifiers

Chemical Name or Material Glycine
CAS 7732-18-5
CAS Max % 93
CAS 56-40-6
CAS Max % 7
Name Note For Ozone
Linear Formula NH2CH2COOH
Formula Weight 75.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC148787

 
lab chem
LC148787
125mL Each for $73.23
Description & Specifications

Specifications

Formula Weight 75.07
Chemical Name or Material Glycine
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 125mL
Name Note For Ozone
Physical Form Liquid
Assay 7% (w/v)
Linear Formula NH2CH2COOH
CAS 7732-18-5
CAS Max % 93
Health Hazard 2 GHS H Statement
Product is not hazardous.
Identification Passes Test
CAS 56-40-6
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 7
Solubility Soluble in water
Storage Information Refrigerate
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.