Please login to your online account to display your discounted pricing

Glycine, For Chlorine Dioxide, 10% (w/v), LabChem™

$69.26 - $77.54

Chemical Identifiers

Chemical Name or Material Glycine
CAS 7732-18-5
CAS Max % 90
CAS 56-40-6
CAS Max % 10
Name Note For Chlorine Dioxide
Linear Formula NH2CH2COOH
Formula Weight 75.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC148802

 
lab chem
LC148802
1L Each for $77.54

LC148801

 
lab chem
LC148801
500mL Each for $69.26
Description & Specifications

Specifications

Formula Weight 75.07
Chemical Name or Material Glycine
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 1L
Name Note For Chlorine Dioxide
Physical Form Liquid
Assay 10% (w/v)
Linear Formula NH2CH2COOH
CAS 7732-18-5
CAS Max % 90
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 56-40-6
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 10
Solubility Soluble in water
Storage Information Refrigerate
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.