missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycine, For Chlorine Dioxide, 10% (w/v), LabChem™
Supplier: LabChem LC148802
Specifications
Glycine | |
For Chlorine Dioxide | |
90,10 | |
NH2CH2COOH | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
750 | |
75.07 | |
Passes Test | |
Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
Liquid |
Colorless | |
56-40-6,7732-18-5 | |
C2H5NO2 | |
MFCD00008131 | |
Soluble in water | |
NCC(O)=O | |
75.07 | |
CHEBI:15428 | |
10% (w/v) | |
Poly Bottle | |
1 L |
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Refrigerate