Please login to your online account to display your discounted pricing

Glycine, For Chlorine, Chlorine Dioxide, 20% (w/v), LabChem™

$86.40 - $86.40

Chemical Identifiers

Chemical Name or Material Glycine
CAS 7732-18-5
CAS Max % 80
CAS 56-40-6
CAS Max % 20
Name Note For Chlorine, Chlorine Dioxide
Linear Formula NH2CH2COOH
Formula Weight 75.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC148851

 
lab chem
LC148851
Poly Bottle 500mL Each for $86.40
Description & Specifications

Specifications

Formula Weight 75.07
CAS 56-40-6
Recommended Storage Refrigerate
Packaging Poly Bottle
CAS Max % 80
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Physical Form Liquid
CAS 7732-18-5
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Identification Passes Test
CAS Max % 20
Chemical Name or Material Glycine
Color Clear, colorless to slight yellow
Assay 20% (w/v)
Quantity 500mL
Solubility Information Soluble in water
Linear Formula NH2CH2COOH
Name Note For Chlorine, Chlorine Dioxide
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Refrigerate