Please login to your online account to display your discounted pricing

Glycine, Crystalline, MP Biomedicals


Quantity 5kg

Manufacturer: mp biomedicals  0219468105

 View more versions of this product

Catalog No. ICN19468105

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5kg
Chemical Name or Material Glycine
Name Note Free Acid
CAS 56-40-6
Purity Grade Cell Culture Reagent
Physical Form Crystalline
Molecular Formula C2H5NO2
Formula Weight 75.07g/mol
Synonym Aminoacetic acid
Recommended Storage Room temperature

Specifications

Chemical Name or Material Glycine
Name Note Free Acid
CAS 56-40-6
Purity Grade Cell Culture Reagent
Physical Form Crystalline
Molecular Formula C2H5NO2
Formula Weight 75.07g/mol
Synonym Aminoacetic acid
Accessories

Room temperature