Please login to your online account to display your discounted pricing

Glycine, ACS Grade, ≥ 98.5%, LabChem™

$114.26 - $114.26

Chemical Identifiers

Chemical Name or Material Glycine
CAS 56-40-6
CAS Max % 100
Purity Grade ACS Grade
Linear Formula NH2CH2COOH
Formula Weight 75.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC148751

 
lab chem
LC148751
Poly Bottle 500g Each for $114.26
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 75.07
Chemical Name or Material Glycine
EINECS Number 200-272-2
Purity Grade ACS Grade
Physical Form Powder
Identification Passes Test
Linear Formula NH2CH2COOH
CAS 56-40-6
Packaging Poly Bottle
Solubility Soluble in water
CAS Max % 100
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Density 1.1607g/cm3
Color White
Melting Point 182°C
Quantity 500g
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Flash Point 176.67°C
Specific Gravity 1.1607g/cm3
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature