Learn More
Glycinamide hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 120061000
Specifications
Glycinamide hydrochloride | |
1% max. (105°C, 4 hrs) | |
Beige to White | |
100g | |
1668-10-6 | |
C2H6N2O·HCl | |
MFCD00013008 | |
2-aminoacetamide hydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamid hydrochloride, glycinamide hcl, glycinamide hydrochloride, glycinamidehydrochloride, glycine amide hydrochloride, glycineamide hcl, glycineamide hydrochloride, h-gly-nh2.hcl | |
C(C(=O)N)N.Cl | |
110.54 | |
110.54 | |
98% |
Authentic | |
Plastic bottle | |
204°C | |
98% | |
97.5% min. (Argentometry) | |
H2NCH2CONH2·HCl | |
04, 343 | |
WKNMKGVLOWGGOU-UHFFFAOYSA-N | |
2-aminoacetamide;hydrochloride | |
2723639 | |
Crystalline Powder |
Chemical Identifiers
1668-10-6 | |
110.54 | |
WKNMKGVLOWGGOU-UHFFFAOYSA-N | |
2723639 | |
C(C(=O)N)N.Cl |
C2H6N2O·HCl | |
MFCD00013008 | |
2-aminoacetamide hydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamid hydrochloride, glycinamide hcl, glycinamide hydrochloride, glycinamidehydrochloride, glycine amide hydrochloride, glycineamide hcl, glycineamide hydrochloride, h-gly-nh2.hcl | |
2-aminoacetamide;hydrochloride |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 216-789-1