missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycidyl Phenyl Ether 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC135350010

 View more versions of this product

Catalog No. AC135350010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Glycidyl phenyl ether
122-60-1
Liquid
99%
C9H10O2
MFCD00005133
1, 2-Epoxy-3-phenoxypropane
Clear colorless to yellow
99%
Authentic
≥98.5%
Glass bottle
150.18
1.5295 to 1.5315 (20°C, 589nm)
1.1000g/mL
1kg

Chemical Identifiers

122-60-1
C9H10O2
MFCD00005133
Glycidyl phenyl ether, 99%
99%
150.18
1, 2-Epoxy-3-phenoxypropane
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-557-2

RTECSNumber : TZ3675000

TSCA : TSCA