missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycidyl Methacrylate, Stabilized 97%, ACROS Organics™

Manufacturer:  Acros Organics 165890050

 View more versions of this product

Catalog No. AC165890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Glycidyl methacrylate
76°C
Glass bottle
50 to 150ppm MEHQ
189.0°C
-41.0°C
106-91-2,150-76-5
C7H10O3
Solubility in water: insoluble. Other solubilities: miscible in almost all common organic solvents
CC(=C)C(=O)OCC1CO1
142.154
142.15
97%
1.0700g/mL
Authentic
1.4490 to 1.4510
0.1% Max. (K.F.)
Undesignated
5g
97%
glycidyl methacrylate, 2,3-epoxypropyl methacrylate, sy-monomer g, glycidol methacrylate, acriester g, blemmer g, blemmer gma, light ester g, glycidylmethacrylate, 2-methacryloxy methyl oxirane
VOZRXNHHFUQHIL-UHFFFAOYSA-N
oxiran-2-ylmethyl 2-methylprop-2-enoate
7837
Liquid

Chemical Identifiers

106-91-2
142.154
glycidyl methacrylate, 2,3-epoxypropyl methacrylate, sy-monomer g, glycidol methacrylate, acriester g, blemmer g, blemmer gma, light ester g, glycidylmethacrylate, 2-methacryloxy methyl oxirane
oxiran-2-ylmethyl 2-methylprop-2-enoate
C7H10O3
VOZRXNHHFUQHIL-UHFFFAOYSA-N
7837
CC(=C)C(=O)OCC1CO1
Safety and Handling

Safety and Handling

GHS P Statement May cause an allergic skin reaction. Causes severe skin burns and eye damage. Harmful if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May damage fertility.

GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

SDS Documents