Please login to your online account to display your discounted pricing

Glycidyl Methacrylate, Stabilized 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC165895000

 View more versions of this product

Catalog No. AC165895000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0% (GC)
Formula Weight 142.15
Quantity 500g
Color Clear
Water 0.1% Max. (K.F.)
Density 1.0700g/mL
Chemical Name or Material Glycidyl methacrylate
Infrared Spectrum Authentic
Molecular Formula C7H10O3
CAS 106-91-2
Stabilizer 50 to 150ppm MEHQ
CAS 150-76-5
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4490 to 1.4510 (20°C, 589nm)
Synonym 2, 3-Epoxypropyl methacrylate
Health Hazard 2 GHS P Statement May cause an allergic skin reaction. Causes severe skin burns and eye damage. Harmful if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May damage fertility.
Health Hazard 3 GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Specifications

Chemical Name or Material Glycidyl methacrylate
CAS 106-91-2
CAS 150-76-5
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C7H10O3
Formula Weight 142.15
Synonym 2, 3-Epoxypropyl methacrylate
Accessories

GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

GHS P Statement May cause an allergic skin reaction. Causes severe skin burns and eye damage. Harmful if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May damage fertility.

SDS & Safety Documents