Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycidyl 4-methoxyphenyl ether, 98%

$69.89 - $604.93

Chemical Identifiers

Chemical Name or Material Glycidyl 4-methoxyphenyl ether
CAS 2211-94-1
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00005135
Synonym 2,3-Epoxypropyl 4-methoxyphenyl ether; 4-(Glycidyloxy)anisole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1978006

  SDS
alfa aesar™
A1978006
5g Each for $69.89

AAA1978022

  SDS
alfa aesar™
A1978022
100g Each for $604.93
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material Glycidyl 4-methoxyphenyl ether
Assay 98%
Molecular Formula C10H12O3
MDL Number MFCD00005135
CAS 2211-94-1
Synonym 2,3-Epoxypropyl 4-methoxyphenyl ether; 4-(Glycidyloxy)anisole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 45° to 48°C
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DA0176000
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.