missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycidyl 2-Methoxyphenyl Ether 98.0+%, TCI America™
$81.29 - $325.16
Chemical Identifiers
CAS | 2210-74-4 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.203 |
MDL Number | MFCD00190158 |
InChI Key | RJNVSQLNEALZLC-UHFFFAOYSA-N |
Synonym | 1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german |
PubChem CID | 98357 |
IUPAC Name | 2-[(2-methoxyphenoxy)methyl]oxirane |
SMILES | COC1=CC=CC=C1OCC2CO2 |
Chemical Identifiers
2210-74-4 | |
180.203 | |
RJNVSQLNEALZLC-UHFFFAOYSA-N | |
98357 | |
COC1=CC=CC=C1OCC2CO2 |
C10H12O3 | |
MFCD00190158 | |
1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german | |
2-[(2-methoxyphenoxy)methyl]oxirane |
Specifications
2210-74-4 | |
White | |
C10H12O3 | |
5 g | |
RJNVSQLNEALZLC-UHFFFAOYSA-N | |
2-[(2-methoxyphenoxy)methyl]oxirane | |
98357 | |
≥98.0% (GC) | |
Glycidyl 2-Methoxyphenyl Ether |
39°C | |
120°C | |
MFCD00190158 | |
1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german | |
COC1=CC=CC=C1OCC2CO2 | |
180.203 | |
180.20 | |
Crystalline Lumps |
Safety and Handling
RTECSNumber : TZ3550000
TSCA : No
Recommended Storage : Refrigerator