Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycidyl 2-methoxyphenyl ether, 98%

Chemical Identifiers

Chemical Name or Material Glycidyl 2-methoxyphenyl ether
CAS 2210-74-4
Formula Weight 180.2
MDL Number MFCD00190158
Synonym Guaiacol glycidyl ether; 2-[(2-Methoxyphenoxy)methyl]oxirane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6177706

  SDS
alfa aesar™
H6177706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6177714

  SDS
alfa aesar™
H6177714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 152° to 158°C
Formula Weight 180.2
Melting Point 41° to 48°C
Physical Form Solid
CAS 2210-74-4
Color Off-white
Assay 98%
Chemical Name or Material Glycidyl 2-methoxyphenyl ether
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number TZ3550000
MDL Number MFCD00190158
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 5g
Synonym Guaiacol glycidyl ether; 2-[(2-Methoxyphenoxy)methyl]oxirane
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.