missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Glycidyl 2-methoxyphenyl ether, 98%

Manufacturer:  Alfa Aesar™ H6177714

 View more versions of this product

Catalog No. AAH6177714



Specifications

Specifications

Glycidyl 2-methoxyphenyl ether
White
25g
C10H12O3
1,2-epoxy-3-2-methoxyphenoxy propane
COC1=CC=CC=C1OCC2CO2
180.203
180.2
98%
152°C to 158°C
41°C to 48°C
2210-74-4
MFCD00190158
RJNVSQLNEALZLC-UHFFFAOYSA-N
2-[(2-methoxyphenoxy)methyl]oxirane
98357
Solid

Chemical Identifiers

2210-74-4
180.203
RJNVSQLNEALZLC-UHFFFAOYSA-N
98357
COC1=CC=CC=C1OCC2CO2
C10H12O3
MFCD00190158
1,2-epoxy-3-2-methoxyphenoxy propane
2-[(2-methoxyphenoxy)methyl]oxirane
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TZ3550000

Documents