missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol, ultrapure, Spectrophotometric Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 32450K7

 View more versions of this product

Catalog No. AA32450K7



Specifications

Specifications

Colorless viscous liquid
1.26g/cm3
400°C (752°F)
<0.1 hPa (0mm Hg) at 20°C
Colorless
4L
Sprectrophotometric
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
Glycerol
160°C
Odorless
182°C
18°C
56-81-5
C3H8O3
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
C(C(CO)O)O
92.094
CHEBI:17754
Viscous Liquid

Chemical Identifiers

56-81-5
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
C(C(CO)O)O
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

EINECSNumber : 200-289-5

SDS Documents