missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol (Spectranalyzed™), Fisher Chemical

For use in the ultraviolet range

Manufacturer:  Fisher Chemical G1531

 View more versions of this product

Catalog No. G153-1




Specifications

Specifications

Pass Test
>290°C
160°C
0.005% max.
0.003mbar at 50°C
290°C
18°C
5
Also meets ACS specifications
C3H8O3
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
≥99.5%
Glycerol
1.261g/cm³
Pass Test
Glass Bottle
0.5% max.
Undesignated
1L
56-81-5
≥99.5 %
HOCH2CH(OH)CH2OH
1,2,3-propanetriol
C(C(CO)O)O
92.094
CHEBI:17754
Viscous Liquid
Spectranalyzed™

Chemical Identifiers

56-81-5
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
1,2,3-propanetriol
CHEBI:17754
C(C(CO)O)O
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:1
Instability:0

Documents