Please login to your online account to display your discounted pricing

Glycerol (Spectranalyzed™), Fisher Chemical

For use in the ultraviolet range

$803.25 - $2,767.17

Chemical Identifiers

Chemical Name or Material Glycerol
CAS 56-81-5
Purity Grade Spectranalyzed™
Purity Grade Notes Also meets ACS specifications
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula C3H8O3
Linear Formula HOCH2CH(OH)CH2OH
Formula Weight 92.09
MDL Number MFCD00004722
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

G153-4

  Accessories
fisher scientific company
BPG1534
Glass Bottle 4L Each for $2,767.17

G153-1

 
fisher scientific company
G1531
Glass Bottle 1L Each for $803.25
Case of 6 Each for $4,819.50
Description & Specifications

Specifications

Assay ≥99.5%
Physical Form Viscous Liquid
Formula Weight 92.09
Boiling Point 290°C
Melting Point 18°C
Quantity 4L
Flash Point 160°C
pH 5
Color Clear
Vapor Pressure 0.003mbar at 50°C
Purity Grade Spectranalyzed™
Density 1.261g/cm³
Decomposition Information >290°C
Fatty Acids and Esters 0.05% max.
Heavy Metals (as Pb) 2ppm max.
Assay Percent Range ≥99.5 %
Acrolein Glucose and NH4 Pass Test
Identification Pass Test
Water 0.5% max.
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:1
Instability:0
ChemAlert Storage Symbol Gray
Sulfate 0.001% max.
Packaging Glass Bottle
Additional Information Vapor Pressure: 0.003mbar at 50°C
Chlorine Compounds 0.003% max.
Neutrality Pass Test
Absorbance Pass Test
Substances Darkened by H2SO4 Pass Test
Synonym 1, 2, 3-Propanetriol
Ignition Residue 0.005% max.
MDL Number MFCD00004722
Linear Formula HOCH2CH(OH)CH2OH
CAS 56-81-5
Molecular Formula C3H8O3
Purity Grade Notes Also meets ACS specifications
Chemical Name or Material Glycerol

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:1
Instability:0

WARNING!