missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol, Reagent ACS, 99.6%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 410985000

 View more versions of this product

Catalog No. AC410985000


 

Specifications Description & Specifications

Specifications

≥99.5%
ACS Reagent Grade
500mL
18.0°C
Clear
1.2610g/mL
01, 502
Passes Test
0.003% max.
10ppm max.
0.5% max. (K.F.)
Passes Test
Vapor Pressure: 0.003mbar at 50°C
2ppm max.
C3H8O3
99.6%
92.09
Liquid
160°C
290.0°C
EcoSafPak™ Glass Bottle
>290°C
15, 4520
0.005% max.
Glycerine
MFCD00004722
1.4700 to 1.4750 (20°C, 589nm)
0.05% max. (as butyric acid)
Authentic
56-81-5
Glycerol
HOCH2CH(OH)CH2OH

Chemical Identifiers

56-81-5
C3H8O3
92.09
Glycerine
≥99.5%
99.6%
HOCH2CH(OH)CH2OH
MFCD00004722
Glycerol
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

HYGROSCOPIC

EINECSNumber : 200-289-5

RTECSNumber : MA8050000

TSCA : TSCA