Please login to your online account to display your discounted pricing

Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents


Quantity 5mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP6455

 View more versions of this product

Catalog No. BP645-5

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Quantity 5mL
Physical Form Liquid
DNase DNase free
Color Colorless
Chemical Name or Material Glycerol Gel-Loading Dye, 5X
CAS 7732-18-5
Absorbance 0.25 to 0.40 AU
Name Note Contains 30% Glycerol
Electrophoresis Pass Test
CAS 56-81-5
ChemAlert Storage Symbol Gray
Concentration or Composition (by Analyte or Components) Glycerol (30%), Proprietary Components (<6%), and Water
Packaging Poly Bottle
Recommended Storage RT

Specifications

Chemical Name or Material Glycerol Gel-Loading Dye, 5X
Name Note Contains 30% Glycerol
CAS 56-81-5
CAS 7732-18-5
Purity Grade Molecular Biology Grade
Accessories
 

RT

SDS & Safety Documents