missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents

Manufacturer:  Fisher Scientific Company FLBP6451

 View more versions of this product

Catalog No. BP645-1


 

Specifications Description & Specifications

Specifications

Molecular Biology Grade
Liquid
DNase free
0.25 to 0.40 AU
Pass Test
Glycerol (30%), Proprietary Components (<6%), and Water
Contains 30% Glycerol
1mL
Colorless
>1.0g/mol
56-81-5,7732-18-5
Poly Tube
Glycerol Gel-Loading Dye, 5X

Chemical Identifiers

56-81-5,7732-18-5
Glycerol Gel-Loading Dye, 5X, Contains 30% Glycerol
Molecular Biology Grade

Safety and Handling

SDS & Safety Documents

Recommended Storage : RT