missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol formal, 99+%, mixture of isomers, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 259775000

 View more versions of this product

Catalog No. AC259775000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Glycerol formal
97°C
Glass bottle
1.21
0.5% max. (K.F.)
Undesignated
4.0 to 6.5 (10% soln.)
4740-78-7,5464-28-8
C4H8O3
14, 4485
104.11
Liquid
1.2100g/mL
Authentic
1.4500 to 1.4520
16 mPa.s (20°C)
193.0°C to 195.0°C
500mL
99+%
99% min. Sum of alpha-and beta-isomers (GC)
MFCD00003218
(50% in water) Clear
104.11
99+%
SDS
Documents