missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycerol formal, 99+%, mixture of isomers, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 259772500
Specifications
Glycerol formal | |
97°C | |
Glass bottle | |
1.21 | |
0.5% max. (K.F.) | |
Undesignated | |
4.0 to 6.5 (10% soln.) | |
4740-78-7,5464-28-8 | |
C4H8O3 | |
14, 4485 | |
104.11 | |
Liquid |
1.2100g/mL | |
Authentic | |
1.4500 to 1.4520 | |
16 mPa.s (20°C) | |
193.0°C to 195.0°C | |
250mL | |
99+% | |
99% min. Sum of alpha-and beta-isomers (GC) | |
MFCD00003218 | |
(50% in water) Clear | |
104.11 | |
99+% |