missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol (Clear, Colorless/USP/FCC), Fisher Chemical

Manufacturer:  Fisher Chemical G31500

 View more versions of this product

Catalog No. G31-500




Specifications

Specifications

Glycerol
1.261g/cm³
Pass Test (FCC/USP)
Glass Bottle
1.249 min. (FCC/USP)
5.0% max. (USP)
Undesignated
500mL
Clear, Colorless
C3H8O3
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.1
99.0 to 101.0% (USP)
>290°C
160°C
0.01% max. (FCC/USP)
Meets Requirements (USP)
0.003mbar at 50°C
290°C
18°C
5
56-81-5
HOCH2CH(OH)CH2OH
1,2,3-propanetriol
C(C(CO)O)O
92.094
CHEBI:17754
Viscous Liquid
FCC/USP

Chemical Identifiers

56-81-5
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
1,2,3-propanetriol
CHEBI:17754
C(C(CO)O)O
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:1
Instability:0

Documents