Please login to your online account to display your discounted pricing

Glycerol, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$544.03 - $544.03

Chemical Identifiers

Chemical Name or Material Glycerol
CAS 56-81-5
Molecular Formula HOCH2CHOHCH2OH
Formula Weight 92.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002932

 
avantor performance
M77809
Narrow Mouth Amber Glass Bottle 4L Each for $544.03
Description & Specifications

Specifications

Formula Weight 92.1
Flash Point 199.00°C
Chemical Name or Material Glycerol
CAS 56-81-5
Molecular Formula HOCH2CHOHCH2OH
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.26kg
Quantity 4L