missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycerol, for analysis, 86-88% wt% aqueous solution, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 295600100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Glycerol | |
56-81-5,7732-18-5 | |
290.0°C | |
Authentic | |
C3H8O3 | |
HOCH2CH(OH)CH2OH | |
10L | |
1.227 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
CHEBI:17754 | |
86.0 to 88.0% |
86 to 88% wt% Aqueous Solution | |
1.2270g/mL | |
160°C | |
Plastic drum | |
1.4520 to 1.4750 | |
MFCD00004722 | |
01, 502 | |
15, 4520 | |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform and in, fixed and volatile oils | |
OCC(O)CO | |
92.09 | |
1069 mPa.s (20°C) | |
92.09 | |
Analysis |
Safety and Handling
EINECSNumber : 200-289-5
RUO â Research Use Only